Product Details

SNP ID
rs76169194
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:207110296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGTAGATGATGTGGACAGGAA[C/T]GGGAGATAGGAAAGTGCTGGGAATG
Phenotype
MIM: 120830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4BPA PubMed Links

Gene Details

Gene
C4BPA
Gene Name
complement component 4 binding protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000715.3 Intron NP_000706.1
XM_005273251.1 Intron XP_005273308.1
XM_005273252.4 Intron XP_005273309.1

View Full Product Details