Product Details

SNP ID
rs75015159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60518045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGTTTTAAATTACATTATTTC[A/G]GTACTAACATAATTCTCTTCTCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A13 PubMed Links

Gene Details

Gene
MS4A13
Gene Name
membrane spanning 4-domains A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012417.2 Intron NP_001012417.2
NM_001100909.1 Intron NP_001094379.1
NM_001278320.1 Intron NP_001265249.1

View Full Product Details