Product Details

SNP ID
rs77687368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCTCTCAAAGGCTGCCTCAC[C/T]CAGCTGTTTGTGGAGCATTTCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 291 Silent Mutation ACC,ACT T97T NP_001004704.1

View Full Product Details