Product Details

SNP ID
rs79179693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55796195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTCAATGAGATGTGTACACTA[C/G]TGATCATCCTCACTTCCTATGTTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5D14 PubMed Links

Gene Details

Gene
OR5D14
Gene Name
olfactory receptor family 5 subfamily D member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004735.1 640 Missense Mutation CTG,GTG L214V NP_001004735.1

View Full Product Details