Product Details

SNP ID
rs76256029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64314886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGACCGAGAGATTGTGGTCAC[C/T]ATCAGCTGGGCCAAGAGCATCCCAG
Phenotype
MIM: 601998 MIM: 606583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282450.1 935 Intron NP_001269379.1
NM_001282451.1 935 Silent Mutation ACC,ACT T238T NP_001269380.1
NM_004451.4 935 Silent Mutation ACC,ACT T239T NP_004442.3
XM_017017313.1 935 Silent Mutation ACC,ACT T249T XP_016872802.1
Gene
PRDX5
Gene Name
peroxiredoxin 5
There are no transcripts associated with this gene.

Gene
TRMT112
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details