Product Details

SNP ID
rs111558968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAAGCCATATCTTTTTTATTGA[A/G]TAAATCATCATCTCTGTCTCCATCT
Phenotype
MIM: 603415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 6054 Missense Mutation CTC,TTC L1916F NP_002968.1
XM_005246757.2 6054 Missense Mutation CTC,TTC L1927F XP_005246814.1
XM_011511616.2 6054 Missense Mutation CTC,TTC L1927F XP_011509918.1
XM_011511617.2 6054 Missense Mutation CTC,TTC L1927F XP_011509919.1
XM_011511618.2 6054 Missense Mutation CTC,TTC L1916F XP_011509920.1
XM_011511619.2 6054 Intron XP_011509921.1
XM_017004668.1 6054 Missense Mutation CTC,TTC L1798F XP_016860157.1
XM_017004669.1 6054 Missense Mutation CTC,TTC L1679F XP_016860158.1

View Full Product Details