Product Details

SNP ID
rs117066669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177550966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTATCACGAAGCATGATTGTGT[A/G]TTTTGACATGTTTTCTAACAAGGAC
Phenotype
MIM: 603051
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPS PubMed Links

Gene Details

Gene
AGPS
Gene Name
alkylglycerone phosphate synthase
There are no transcripts associated with this gene.

Gene
TTC30B
Gene Name
tetratricopeptide repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152517.2 1831 Missense Mutation CAC,TAC H600Y NP_689730.2

View Full Product Details