Product Details

SNP ID
rs116304177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181678369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCGTCTGAACGAAGGAGACCAG[A/G]TCTGGGCTTTTGCCTGAGCGCAGGA
Phenotype
MIM: 601724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEUROD1 PubMed Links

Gene Details

Gene
NEUROD1
Gene Name
neuronal differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002500.4 739 Silent Mutation GAC,GAT D164D NP_002491.2

View Full Product Details