Product Details

SNP ID
rs114918839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206939664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTCCTGGAGGGCTGGGATATG[A/C]TAGGTGAGTGTAAAGTGGGAAGAGG
Phenotype
MIM: 609563
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPO PubMed Links

Gene Details

Gene
CPO
Gene Name
carboxypeptidase O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173077.2 6218 Missense Mutation GAT,GCT D22A NP_775100.1
XM_017003372.1 6218 Missense Mutation GAT,GCT D22A XP_016858861.1

View Full Product Details