Product Details
- SNP ID
-
rs112919603
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218425776 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGCGAGGTCCAGGGCAACGAGAG[C/T]GAGGCCTTCCGAGGCTACTTCAAGC
- Phenotype
-
MIM: 193040
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VIL1
PubMed Links
Gene Details
- Gene
- VIL1
- Gene Name
- villin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007127.2 |
377 |
Silent Mutation |
AGC,AGT |
S104S |
NP_009058.2 |
View Full Product Details