Product Details

SNP ID
rs113366352
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223975855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTCGTTGCTTTGCATGGTTTCT[C/T]TTGTATACTCTTCAGGGTTTGTTTA
Phenotype
MIM: 611849 MIM: 177010
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPL44 PubMed Links

Gene Details

Gene
MRPL44
Gene Name
mitochondrial ribosomal protein L44
There are no transcripts associated with this gene.

Gene
SERPINE2
Gene Name
serpin family E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136528.1 1267 UTR 3 NP_001130000.1
NM_001136530.1 1267 UTR 3 NP_001130002.1
NM_006216.3 1267 UTR 3 NP_006207.1
XM_005246641.2 1267 UTR 3 XP_005246698.1
XM_017004329.1 1267 UTR 3 XP_016859818.1
XM_017004330.1 1267 UTR 3 XP_016859819.1
XM_017004331.1 1267 UTR 3 XP_016859820.1
XM_017004332.1 1267 UTR 3 XP_016859821.1

View Full Product Details