Product Details

SNP ID
rs114409792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157319208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTGCAAAAACATTGTGTCAT[C/T]CATGAACATGAAGCATTTACCAACT
Phenotype
MIM: 610072 MIM: 615129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERMN PubMed Links

Gene Details

Gene
ERMN
Gene Name
ermin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009959.2 3051 UTR 3 NP_001009959.1
NM_001304344.1 3051 UTR 3 NP_001291273.1
NM_001304345.1 3051 UTR 3 NP_001291274.1
NM_001304346.1 3051 UTR 3 NP_001291275.1
NM_020711.2 3051 UTR 3 NP_065762.1
Gene
GALNT5
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017003237.1 3051 Intron XP_016858726.1
XM_017003238.1 3051 Intron XP_016858727.1

View Full Product Details