Product Details

SNP ID
rs111922023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113721842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGCACATGAGCATCTGCGGGC[C/T]CTATCCTCTTATAGTAGTTCTTCTT
Phenotype
MIM: 610834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAA50 PubMed Links

Gene Details

Gene
NAA50
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308445.1 775 Missense Mutation GAG,GGG E142G NP_001295374.1
NM_025146.3 775 Missense Mutation GAG,GGG E143G NP_079422.1

View Full Product Details