Product Details
- SNP ID
-
rs114428834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:125453359 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGGGAGGGGAGAAGAAATGGTGG[A/G]GGGAAAGTACAAAGGGTAGAAATCA
- Phenotype
-
MIM: 605931
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX4
PubMed Links
Gene Details
- Gene
- SNX4
- Gene Name
- sorting nexin 4
View Full Product Details