Product Details

SNP ID
rs115255996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131382933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATACATCTTTGAGGCAGTAGCCA[A/C]TAGAGGTTTAGAAGCAGATGTGGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC339874 PubMed Links

Gene Details

Gene
LOC339874
Gene Name
uncharacterized LOC339874
There are no transcripts associated with this gene.

Gene
NUDT16
Gene Name
nudix hydrolase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171905.1 Intron NP_001165376.1
NM_001171906.1 Intron NP_001165377.1
NM_152395.2 Intron NP_689608.2

View Full Product Details