Product Details

SNP ID
rs116645637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:190848367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTGGTGCTGTTGAGCTTTTT[A/G]TTCTACCTCTATGTCTACTTGTTGT
Phenotype
MIM: 614448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMNC PubMed Links

Gene Details

Gene
GMNC
Gene Name
geminin coiled-coil domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146686.2 Intron NP_001140158.1

View Full Product Details