Product Details

SNP ID
rs115159413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5054629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCCCAGTGGCCGTGGTCAGGT[A/G]TTTGGACTTGATCTTATAGGCATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107986255 PubMed Links

Gene Details

Gene
LOC107986255
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
STK32B
Gene Name
serine/threonine kinase 32B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306082.1 Intron NP_001293011.1
NM_018401.2 Intron NP_060871.1
XM_005247982.3 Intron XP_005248039.1
XM_005247983.4 Intron XP_005248040.1
XM_011513497.1 Intron XP_011511799.1
XM_011513501.2 Intron XP_011511803.1
XM_011513504.1 Intron XP_011511806.1
XM_017008374.1 Intron XP_016863863.1
XM_017008375.1 Intron XP_016863864.1
XM_017008376.1 Intron XP_016863865.1
XM_017008377.1 Intron XP_016863866.1

View Full Product Details