Product Details

SNP ID
rs117535467
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15960346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTAAACTTTCATAGAAGGCTC[A/G]GATCAACAAAGCAAAACTTCTACAG
Phenotype
MIM: 607713 MIM: 604365
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FGFBP2 PubMed Links

Gene Details

Gene
FGFBP2
Gene Name
fibroblast growth factor binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031950.3 1065 UTR 3 NP_114156.1
Gene
PROM1
Gene Name
prominin 1
There are no transcripts associated with this gene.

View Full Product Details