Product Details

SNP ID
rs112717667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGAAAGATGAACCTTCTGATG[C/T]GAAACTTTAGAGAAGAGAGAAAAAC
Phenotype
MIM: 604796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 1077 Missense Mutation CAC,CGC H253R NP_062545.1

View Full Product Details