Product Details
- SNP ID
-
rs114479610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:62418914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGGTGTGAAACTGCTGGGTCAT[A/G]GAAGATACAGTCACGAGTTGCTTAA
- Phenotype
-
MIM: 610889
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IPO11
PubMed Links
Gene Details
- Gene
- IPO11
- Gene Name
- importin 11
View Full Product Details