Product Details

SNP ID
rs116319565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4996031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAAATAAGCTTTTGCCACCAC[C/T]GTGCTTGGCTCAGGACAGCAATAGG
Phenotype
MIM: 606117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYRM4-AS1 PubMed Links

Gene Details

Gene
LYRM4-AS1
Gene Name
LYRM4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPP40
Gene Name
ribonuclease P/MRP subunit p40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286132.1 748 Silent Mutation ACA,ACG T248T NP_001273061.1
NM_001286133.1 748 Silent Mutation ACA,ACG T229T NP_001273062.1
NM_006638.3 748 Silent Mutation ACA,ACG T271T NP_006629.2
XM_011514253.2 748 Silent Mutation ACA,ACG T271T XP_011512555.1

View Full Product Details