Product Details

SNP ID
rs117587119
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19721755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAATATGTCATTACAAATATA[A/G]TAATCATGCTAGTCAAATAGTGTTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMEM196 PubMed Links

Gene Details

Gene
TMEM196
Gene Name
transmembrane protein 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152774.3 1861 UTR 3 NP_689987.3
XM_005249692.4 1861 UTR 3 XP_005249749.2
XM_011515249.2 1861 UTR 3 XP_011513551.1
XM_011515250.2 1861 UTR 3 XP_011513552.1
XM_017011928.1 1861 Intron XP_016867417.1
XM_017011929.1 1861 Intron XP_016867418.1

View Full Product Details