Product Details

SNP ID
rs117448714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102434868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTCTTCCCTCCATATCTCCTG[A/G]ACAAGGAGAGCCACCCCACATGTAC
Phenotype
MIM: 610929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100630923 PubMed Links

Gene Details

Gene
LOC100630923
Gene Name
LOC100289561-PRKRIP1 readthrough
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 Intron NP_001119812.1
NM_001271818.1 Intron NP_001258747.1
NM_001271819.1 Intron NP_001258748.1
NM_032831.3 Intron NP_116220.1
Gene
PRKRIP1
Gene Name
PRKR interacting protein 1 (IL11 inducible)
There are no transcripts associated with this gene.

View Full Product Details