Product Details

SNP ID
rs113758232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34366945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAATAAAGAAGGTAGAGGAGT[G/T]GTGTATTAGTCTGTTTTCACACTGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1161 PubMed Links

Gene Details

Gene
KIAA1161
Gene Name
KIAA1161
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020702.4 6166 UTR 3 NP_065753.2
XM_011517966.2 6166 UTR 3 XP_011516268.1
XM_017014930.1 6166 UTR 3 XP_016870419.1

View Full Product Details