Product Details

SNP ID
rs111693331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3218588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACGAAGTAGAGTGTGTCCTGGG[C/T]GCCCAGGGCAGCCTGAGCCACGGAG
Phenotype
MIM: 607232 MIM: 607234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRE PubMed Links

Gene Details

Gene
MRGPRE
Gene Name
MAS related GPR family member E
There are no transcripts associated with this gene.

Gene
MRGPRG
Gene Name
MAS related GPR family member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164377.1 226 Missense Mutation NP_001157849.1
Gene
MRGPRG-AS1
Gene Name
MRGPRG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details