Product Details

SNP ID
rs111277483
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46751220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATGGGCCTCCAGCTCAGACTC[A/G]TTTTTGTTGTCGATCATCGTTAGGT
Phenotype
MIM: 611142
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CKAP5 PubMed Links

Gene Details

Gene
CKAP5
Gene Name
cytoskeleton associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008938.3 5484 Silent Mutation AAC,AAT N1786N NP_001008938.1
NM_014756.3 5484 Silent Mutation AAC,AAT N1726N NP_055571.2
Gene
MIR5582
Gene Name
microRNA 5582
There are no transcripts associated with this gene.

View Full Product Details