Product Details

SNP ID
rs115682932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9853147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATACCTGTAACCATTTTTTGCCA[A/G]TCTTTGAAGTGGCTTTTATGTGTAG
Phenotype
MIM: 603242 MIM: 605029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC2B PubMed Links

Gene Details

Gene
CLEC2B
Gene Name
C-type lectin domain family 2 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005127.2 1260 UTR 3 NP_005118.2
Gene
KLRF1
Gene Name
killer cell lectin like receptor F1
There are no transcripts associated with this gene.

View Full Product Details