Product Details

SNP ID
rs111568993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122273103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTGTTTCTCCTGATCATCACT[A/G]CAAATGTTAATGTGTGAAATCAGAA
Phenotype
MIM: 179838 MIM: 610034
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIP1 PubMed Links

Gene Details

Gene
CLIP1
Gene Name
CAP-Gly domain containing linker protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247997.1 Intron NP_001234926.1
NM_002956.2 Intron NP_002947.1
NM_198240.1 Intron NP_937883.1
XM_005253593.2 Intron XP_005253650.1
XM_006719552.3 Intron XP_006719615.1
XM_017019786.1 Intron XP_016875275.1
XM_017019787.1 Intron XP_016875276.1
XM_017019788.1 Intron XP_016875277.1
XM_017019789.1 Intron XP_016875278.1
XM_017019790.1 Intron XP_016875279.1
XM_017019791.1 Intron XP_016875280.1
XM_017019792.1 Intron XP_016875281.1
Gene
VPS33A
Gene Name
VPS33A, CORVET/HOPS core subunit
There are no transcripts associated with this gene.

View Full Product Details