Product Details

SNP ID
rs116277594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113392639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGAATTCCTCACTGCTGTGAT[G/T]CAGGTCTCTCCTGTCCACCCTGCTC
Phenotype
MIM: 182128
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLBD2 PubMed Links

Gene Details

Gene
PLBD2
Gene Name
phospholipase B domain containing 2
There are no transcripts associated with this gene.

Gene
SDS
Gene Name
serine dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006843.2 1411 UTR 3 NP_006834.2

View Full Product Details