Product Details
- SNP ID
-
rs117421680
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1946611 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTGATGGGCCTGGCAGTCGCCC[A/C]CTCCCAGCCTCACCAGCGGTGTGAT
- Phenotype
-
MIM: 606216
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MSRB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35725974] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSRB1
- Gene Name
- methionine sulfoxide reductase B1
There are no transcripts associated with this gene.
- Gene
- RPL3L
- Gene Name
- ribosomal protein L3 like
View Full Product Details