Product Details

SNP ID
rs114923885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29860585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTGCAGGAAATGCTTACCCTC[A/G]AGGTGTAGTAGATCCGAAGCACCGG
Phenotype
MIM: 605893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDIPT PubMed Links

Gene Details

Gene
CDIPT
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286585.1 706 Missense Mutation TCG,TTG S92L NP_001273514.1
NM_001286586.1 706 Missense Mutation TCG,TTG S72L NP_001273515.1
NM_006319.4 706 Missense Mutation TCG,TTG S137L NP_006310.1
XM_005255038.4 706 Missense Mutation TCG,TTG S161L XP_005255095.1
Gene
CDIPT-AS1
Gene Name
CDIPT antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details