Product Details

SNP ID
rs116104149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4965760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCTATTTTAAATTTTATTTAA[A/G]GCCAGTTGTGGTGGCTCACGCATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC14L5 PubMed Links

Gene Details

Gene
SEC14L5
Gene Name
SEC14 like lipid binding 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014692.1 Intron NP_055507.1

View Full Product Details