Product Details

SNP ID
rs111846078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67230627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGCAGACACGGCGGTGCACTAT[C/T]CTTAGCATGGCAACACGGCGGGCAC
Phenotype
MIM: 606881
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318202.1 3238 Silent Mutation NP_001305131.1
NM_013241.2 3238 Silent Mutation NP_037373.2
XM_006721180.1 3238 Missense Mutation XP_006721243.1
XM_011523043.2 3238 Missense Mutation XP_011521345.1
XM_011523044.1 3238 Missense Mutation XP_011521346.1
XM_011523045.2 3238 Missense Mutation XP_011521347.1
Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
TMEM208
Gene Name
transmembrane protein 208
There are no transcripts associated with this gene.

View Full Product Details