Product Details

SNP ID
rs113468622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4731510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAATGGCTACCTACAGCCTGGC[A/G]AACGAGAGACTACGCGCTCTGGAAG
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
There are no transcripts associated with this gene.

Gene
MED11
Gene Name
mediator complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001683.3 103 Silent Mutation GCA,GCG A7A NP_001001683.1
NM_001305000.1 103 Silent Mutation GCA,GCG A7A NP_001291929.1
Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
There are no transcripts associated with this gene.

View Full Product Details