Product Details

SNP ID
rs116939857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7317913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTAGCACAGGCTTCGCTTTGG[C/T]GGAGGCATGAAATAATCTTCTGCGG
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
There are no transcripts associated with this gene.

Gene
GPS2
Gene Name
G protein pathway suppressor 2
There are no transcripts associated with this gene.

Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005408.1 4081 Silent Mutation CCA,CCG P1358P NP_001005408.1
NM_032442.2 4081 Silent Mutation CCA,CCG P1360P NP_115818.2

View Full Product Details