Product Details

SNP ID
rs116312889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8159659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATTGGGGAGGAGGGTTTTCAGT[A/C]CAACCCCCAGCAGGGGTGAGGCCAG
Phenotype
MIM: 602260 MIM: 185881
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PER1 PubMed Links

Gene Details

Gene
PER1
Gene Name
period circadian clock 1
There are no transcripts associated with this gene.

Gene
VAMP2
Gene Name
vesicle associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014232.2 1973 UTR 3 NP_055047.2
XM_005256775.4 1973 UTR 3 XP_005256832.1

View Full Product Details