Product Details

SNP ID
rs112707484
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8160301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCAAAGACCTTGGGATTCTTA[C/G]GACCAAGTGGGGCCAGTCTCAGAGC
Phenotype
MIM: 602260 MIM: 185881
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PER1 PubMed Links

Gene Details

Gene
PER1
Gene Name
period circadian clock 1
There are no transcripts associated with this gene.

Gene
VAMP2
Gene Name
vesicle associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014232.2 1331 UTR 3 NP_055047.2
XM_005256775.4 1331 UTR 3 XP_005256832.1

View Full Product Details