Product Details

SNP ID
rs112966072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63602388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTGAAACAAGGATAATACCCA[A/C]CTTGAGGTAGTTTATGGTTTATGGC
Phenotype
MIM: 605973 MIM: 612958
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCAF7 PubMed Links

Gene Details

Gene
DCAF7
Gene Name
DDB1 and CUL4 associated factor 7
There are no transcripts associated with this gene.

Gene
TACO1
Gene Name
translational activator of cytochrome c oxidase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016360.3 Intron NP_057444.2

View Full Product Details