Product Details

SNP ID
rs112443384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75874928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATGACCCCCATGCTGACCCAG[C/T]CCTCGATAATCTCCACCTCCCAGTA
Phenotype
MIM: 611041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM47 PubMed Links

Gene Details

Gene
TRIM47
Gene Name
tripartite motif containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033452.2 1499 Missense Mutation GAC,GGC D491G NP_258411.2
XM_005257787.4 1499 Missense Mutation GAC,GGC D253G XP_005257844.1
XM_005257788.4 1499 Missense Mutation GAC,GGC D253G XP_005257845.1
Gene
TRIM65
Gene Name
tripartite motif containing 65
There are no transcripts associated with this gene.

View Full Product Details