Product Details

SNP ID
rs117723754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30385019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTCTGGAAATCTGCATGGTGG[C/T]TCAGTGGTAGCAAGCTATCCTTTTG
Phenotype
MIM: 603102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPD PubMed Links

Gene Details

Gene
CPD
Gene Name
carboxypeptidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199775.1 196 Silent Mutation GGC,GGT G12G NP_001186704.1
NM_001304.4 196 Silent Mutation GGC,GGT G259G NP_001295.2

View Full Product Details