Product Details

SNP ID
rs115282214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35817699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTTTAAATAAGATTTAAAATT[A/C]TTTTTATGTGTTCTAGATTCTGGAA
Phenotype
MIM: 601574
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAF15 PubMed Links

Gene Details

Gene
TAF15
Gene Name
TATA-box binding protein associated factor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003487.3 Intron NP_003478.1
NM_139215.2 Intron NP_631961.1
XM_011525315.1 Intron XP_011523617.1
XM_017025181.1 Intron XP_016880670.1

View Full Product Details