Product Details

SNP ID
rs117476810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:1271754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGAAACTACCCCTAAGTCACA[C/T]CCTTCTCCTTCTTTAGGTGGTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00470 PubMed Links

Gene Details

Gene
LINC00470
Gene Name
long intergenic non-protein coding RNA 470
There are no transcripts associated with this gene.

View Full Product Details