Product Details

SNP ID
rs112950542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5891612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCAGAGAAGATGCGGAAGAAG[A/C]AGCCCACGGACGTGGAGGAGGAGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 493 Missense Mutation TGC,TTC C151F NP_001073678.1
XM_005258130.4 493 Missense Mutation TGC,TTC C151F XP_005258187.1
XM_011525729.2 493 Missense Mutation TGC,TTC C151F XP_011524031.1

View Full Product Details