Product Details

SNP ID
rs114790937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24430352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTTAAAGGGCAAGAACGTGC[A/G]GATTTATCAAATAGCCTTGAGGAAA
Phenotype
MIM: 615319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IMPACT PubMed Links

Gene Details

Gene
IMPACT
Gene Name
impact RWD domain protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018439.3 361 Silent Mutation GCA,GCG A83A NP_060909.1
XM_017025832.1 361 Silent Mutation GCA,GCG A65A XP_016881321.1

View Full Product Details