Product Details

SNP ID
rs111552641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40441979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGACTTTGTCTAGGGAGATGCG[A/G]AGCAGGGCTTGCTGGTAGCTCTGAA
Phenotype
MIM: 600941 MIM: 612125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLVRB PubMed Links

Gene Details

Gene
BLVRB
Gene Name
biliverdin reductase B
There are no transcripts associated with this gene.

Gene
LOC105372489
Gene Name
uncharacterized LOC105372489
There are no transcripts associated with this gene.

Gene
SERTAD3
Gene Name
SERTA domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013368.3 380 Silent Mutation CTC,CTT L34L NP_037500.2
NM_203344.2 380 Silent Mutation CTC,CTT L34L NP_976219.1
XM_005258832.3 380 Silent Mutation CTC,CTT L34L XP_005258889.1
XM_006723179.3 380 Silent Mutation CTC,CTT L34L XP_006723242.1

View Full Product Details