Product Details
- SNP ID
-
rs116767543
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:42763941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATATAAGGTGAAGGTGAAATGTCC[A/G]GTTACTCCTCTATTCTCATCACCTC
- Phenotype
-
MIM: 176397
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PSG8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7255164] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSG8
- Gene Name
- pregnancy specific beta-1-glycoprotein 8
View Full Product Details