Product Details

SNP ID
rs111412352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44057529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAGTAATACCAGCTAATGTTC[A/C]TTAAACAGTCTGTGCCGTAGACTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF223 PubMed Links

Gene Details

Gene
ZNF223
Gene Name
zinc finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013361.5 Intron NP_037493.3
XM_017027258.1 Intron XP_016882747.1
XM_017027259.1 Intron XP_016882748.1
XM_017027260.1 Intron XP_016882749.1

View Full Product Details