Product Details

SNP ID
rs116333771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44959790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTTACTTTCCCCCCTAGCAGC[A/G]TACGAGGGTTCCTGTTTTTCCACAT
Phenotype
MIM: 604783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1 PubMed Links

Gene Details

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282175.1 Intron NP_001269104.1
NM_001282176.1 Intron NP_001269105.1
NM_001294.3 Intron NP_001285.1

View Full Product Details