Product Details

SNP ID
rs113724718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50963304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCAGCCTCCCACTACTGACCA[C/T]CTGCTGTCTTGCCCCAGCCCAGGAT
Phenotype
MIM: 602652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK6 PubMed Links

Gene Details

Gene
KLK6
Gene Name
kallikrein related peptidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012964.2 884 Missense Mutation GAT,GGT D148G NP_001012982.1
NM_001012965.2 884 Missense Mutation GAT,GGT D41G NP_001012983.1
NM_001319948.1 884 Missense Mutation GAT,GGT D41G NP_001306877.1
NM_001319949.1 884 Missense Mutation GAT,GGT D41G NP_001306878.1
NM_002774.3 884 Missense Mutation GAT,GGT D148G NP_002765.1

View Full Product Details