Product Details

SNP ID
rs111492869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50997777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACGTGTCAGCCCCTTTGCTGC[C/T]GCCTGCACAGACCATGCCATCTGTG
Phenotype
MIM: 605644 MIM: 605504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK8 PubMed Links

Gene Details

Gene
KLK8
Gene Name
kallikrein related peptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281431.1 357 Missense Mutation AGC,GGC S80G NP_001268360.1
NM_007196.3 357 Missense Mutation AGC,GGC S201G NP_009127.1
NM_144505.2 357 Missense Mutation AGC,GGC S246G NP_653088.1
NM_144506.2 357 Missense Mutation AGC,GGC S60G NP_653089.1
NM_144507.2 357 Intron NP_653090.1
Gene
KLK9
Gene Name
kallikrein related peptidase 9
There are no transcripts associated with this gene.

View Full Product Details